LIF448693574 Life Chemicals Inc.

1,4-oxazepane hydrochloride

  • MDL number MFCD02683070
  • Cas Number : 178312-62-4
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart