LIF448693577 Life Chemicals Inc.

2-(3-(trifluoromethyl)-1H-pyrazol-1-yl)ethan-1-amine

  • MDL number MFCD04970067
  • Cas Number : 1006436-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart