LIF448693585 Life Chemicals Inc.

6-methyl-1H-imidazo[1,2-b]pyrazole

  • MDL number MFCD18450316
  • Cas Number : 42351-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart