LIF448693589 Life Chemicals Inc.

methyl 2-(4-chloro-6-methoxypyrimidin-5-yl)acetate

  • MDL number MFCD28388809
  • Cas Number : 171096-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart