LIF448693590 Life Chemicals Inc.

5-cyclopropyl-3-(thiophen-2-yl)-1H-pyrazole

  • MDL number MFCD28388810
  • Cas Number : 320718-46-5
  • Molecular Weight : 190.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart