LIF448693596 Life Chemicals Inc.

3-(tert-butyl)-1-(2-methoxyphenyl)-1H-pyrazol-5-amine

  • MDL number MFCD04115093
  • Cas Number : 862368-61-4
  • Molecular Weight : 245.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart