LIF448902233 Life Chemicals Inc.

3-bromo-4-methyl-1,5-naphthyridine

  • MDL number MFCD29037849
  • Cas Number : 1820666-67-8
  • Molecular Weight : 223.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart