LIF448902241 Life Chemicals Inc.

3-(5-methoxy-1,2,4-oxadiazol-3-yl)aniline

  • MDL number MFCD23827055
  • Cas Number : 1538156-84-1
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart