LIF448902247 Life Chemicals Inc.

2-bromo-7-fluoroquinoline

  • MDL number MFCD11042212
  • Cas Number : 181147-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart