LIF448902254 Life Chemicals Inc.

azepane-1-sulfonyl chloride

  • MDL number MFCD08444495
  • Cas Number : 41483-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart