LIF448902265 Life Chemicals Inc.

2-amino-4-chloro-5-methylbenzoic acid

  • MDL number MFCD06658301
  • Cas Number : 155184-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart