LIF448902268 Life Chemicals Inc.

N-methoxy-N-methylnicotinamide

  • MDL number MFCD11847417
  • Cas Number : 95091-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart