LIF455764002 Life Chemicals Inc.

2-oxo-1-(pyridin-2-ylmethyl)-1,2-dihydropyridine-3-carboxylic acid hydrochloride

  • MDL number MFCD29037877
  • Cas Number : 1820684-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart