LIF455764003 Life Chemicals Inc.

3-amino-4,5-dichlorobenzoic acid hydrochloride

  • MDL number MFCD29037850
  • Cas Number : 1820619-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart