LIF455764006 Life Chemicals Inc.

morpholine-2-carbonitrile 4-methylbenzenesulfonate

  • MDL number MFCD29037851
  • Cas Number : 1820717-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart