LIF455764010 Life Chemicals Inc.

4-formyl-N,N-dimethylbenzenesulfonamide

  • MDL number MFCD09817632
  • Cas Number : 54049-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart