LIF455764014 Life Chemicals Inc.

6-methoxy-1,2,3,4-tetrahydro-1,8-naphthyridine

  • MDL number MFCD27921554
  • Cas Number : 1820735-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart