LIF455764017 Life Chemicals Inc.

3-(methylthio)azetidine hydrochloride

  • MDL number MFCD17015916
  • Cas Number : 1417793-19-1
  • Molecular Weight : 139.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart