LIF455764021 Life Chemicals Inc.

(3-methoxybenzyl)hydrazine hydrochloride

  • MDL number MFCD11983253
  • Cas Number : 179108-95-3
  • Molecular Weight : 188.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-methoxybenzyl)hydrazine hydrochloride
Inchi Code 1S/C8H12N2O.ClH/c1-11-8-4-2-3-7(5-8)6-10-9;/h2-5,10H,6,9H2,1H3;1H
Inchi Key NZBMDBPDUKFLCT-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart