LIF455764023 Life Chemicals Inc.

tert-butyl 3,4-dihydroquinoxaline-1(2H)-carboxylate

  • MDL number MFCD06656598
  • Cas Number : 887590-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart