LIF455764025 Life Chemicals Inc.

2,3-dihydro-1-benzothiophen-3-one

  • MDL number MFCD08061607
  • Cas Number : 130-03-0
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-benzothiophen-3(2H)-one
Inchi Code 1S/C8H6OS/c9-7-5-10-8-4-2-1-3-6(7)8/h1-5,9H
Inchi Key XHQBIYCRFVVHFD-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart