LIF455764026 Life Chemicals Inc.

(R)-isoxazolidin-4-ol hydrochloride

  • MDL number MFCD20483792
  • Cas Number : 338464-48-5
  • Molecular Weight : 125.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-isoxazolidin-4-ol hydrochloride
Inchi Code 1S/C3H7NO2.ClH/c5-3-1-4-6-2-3;/h3-5H,1-2H2;1H/t3-;/m1./s1
Inchi Key LLCWGQYEQKYRDO-AENDTGMFSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart