LIF455764037 Life Chemicals Inc.

1-(bromomethyl)cyclohexan-1-ol

  • MDL number MFCD20622090
  • Cas Number : 17299-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart