LIF455764046 Life Chemicals Inc.

ethyl 4-cyclopropyl-3-oxobutanoate

  • MDL number MFCD06656674
  • Cas Number : 630399-84-7
  • Molecular Weight : 170.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart