LIF457016090 Life Chemicals Inc.

2-azaspiro[3.5]nonane hydrobromide

  • MDL number MFCD29037837
  • Cas Number : 1820706-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart