LIF457016092 Life Chemicals Inc.

1-(6-phenylpyridin-3-yl)methanamine hydrochloride

  • MDL number MFCD29046741
  • Cas Number : 2034155-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart