LIF457016105 Life Chemicals Inc.

methylene bis(dimethylcarbamodithioate)

  • MDL number MFCD00790886
  • Cas Number : 22656-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart