LIF457016110 Life Chemicals Inc.

octahydrocyclopenta[b]morpholine hydrochloride

  • MDL number MFCD13195915
  • Cas Number : 1197767-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart