LIF457016112 Life Chemicals Inc.

3-bromophenylthiourea

  • MDL number MFCD00060440
  • Cas Number : 21327-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart