LIF457016122 Life Chemicals Inc.

3-amino-1,1,1-trifluoropropan-2-ol hydrochloride

  • MDL number MFCD07784296
  • Cas Number : 3832-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart