LIF457016124 Life Chemicals Inc.

5-(aminomethyl)pyrrolidin-2-one hydrochloride

  • MDL number MFCD09701392
  • Cas Number : 115307-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart