LIF457016131 Life Chemicals Inc.

tert-butyl 3-(4-aminophenoxy)piperidine-1-carboxylate

  • MDL number MFCD06658353
  • Cas Number : 643087-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart