LIF458786925 Life Chemicals Inc.

1-(3-aminopropyl)azepan-2-one oxalate

  • MDL number MFCD28369742
  • Molecular Weight : 260.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart