LIF458786926 Life Chemicals Inc.

2-(1-methyl-1H-pyrazol-5-yl)cyclopropan-1-amine hydrochloride

  • MDL number MFCD29038158
  • Cas Number : 1820711-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart