LIF458786929 Life Chemicals Inc.

6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,3]dioxolo[4,5-b]pyridine

  • MDL number MFCD29038168

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart