LIF458786931 Life Chemicals Inc.

4-methyl-1H-imidazole-5-carboxylic acid hydrochloride

  • MDL number MFCD28398174
  • Cas Number : 82616-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart