LIF458786933 Life Chemicals Inc.

5,6,7,8-tetrahydro-1,8-naphthyridin-3-ol hydrochloride

  • MDL number MFCD29038155
  • Cas Number : 1820665-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart