LIF458786935 Life Chemicals Inc.

6-hydroxy-3H-imidazo[4,5-b]pyridine-5-carboxylic acid

  • MDL number MFCD29038166
  • Cas Number : 1820703-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart