LIF458786936 Life Chemicals Inc.

6-methoxy-3H-imidazo[4,5-b]pyridine-7-carboxylic acid

  • MDL number MFCD29038165
  • Cas Number : 1820717-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart