LIF458786939 Life Chemicals Inc.

2,7-dimethoxy-1,8-naphthyridine-4-carbaldehyde

  • MDL number MFCD29038162
  • Cas Number : 1820648-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart