LIF458786942 Life Chemicals Inc.

2-(2-cyclopropyl-1H-imidazol-1-yl)acetic acid hydrochloride

  • MDL number MFCD29038153
  • Cas Number : 1949816-62-9
  • Molecular Weight : 202.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart