LIF458786943 Life Chemicals Inc.

4,5,6,7-tetrahydropyrazolo[1,5-a]pyridin-5-amine dihydrochloride

  • MDL number MFCD29046921
  • Cas Number : 2034157-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart