LIF458786945 Life Chemicals Inc.

N-methyl-2-(1,3,5-trimethyl-1H-pyrazol-4-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD29038152
  • Cas Number : 1820706-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart