LIF458786951 Life Chemicals Inc.

4-((4-chlorobenzylidene)amino)benzenesulfonamide

  • MDL number MFCD00567080
  • Cas Number : 1268249-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart