LIF458786952 Life Chemicals Inc.

(4-methyl-1H-imidazol-5-yl)methanol

  • MDL number MFCD00072702
  • Cas Number : 29636-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart