LIF458786953 Life Chemicals Inc.

tert-butyl hexahydropyrrolo[3,4-b]pyrrole-1(2H)-carboxylate

  • MDL number MFCD06739059
  • Cas Number : 185693-02-1
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl hexahydropyrrolo[3,4-b]pyrrole-1(2H)-carboxylate
Inchi Code 1S/C11H20N2O2/c1-11(2,3)15-10(14)13-5-4-8-6-12-7-9(8)13/h8-9,12H,4-7H2,1-3H3
Inchi Key OIKZNKIPPFFTKA-UHFFFAOYSA-N

Description

Purity 95%
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart