LIF458786955 Life Chemicals Inc.

2-benzyloctahydro-4H-isoindol-4-one

  • MDL number MFCD10758102
  • Cas Number : 879687-90-8
  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-benzyloctahydro-4H-isoindol-4-one
Inchi Code 1S/C15H19NO/c17-15-8-4-7-13-10-16(11-14(13)15)9-12-5-2-1-3-6-12/h1-3,5-6,13-14H,4,7-11H2
Inchi Key CEOKBKQHDGSNSQ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart