LIF458786956 Life Chemicals Inc.

indoline-6-carboxylic acid hydrochloride

  • MDL number MFCD09878906
  • Cas Number : 15861-37-7
  • Molecular Weight : 199.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-indolinecarboxylic acid hydrochloride
Inchi Code 1S/C9H9NO2.ClH/c11-9(12)7-2-1-6-3-4-10-8(6)5-7;/h1-2,5,10H,3-4H2,(H,11,12);1H
Inchi Key GAPSTRMYKHJESC-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart