LIF458786957 Life Chemicals Inc.

methyl indoline-6-carboxylate hydrochloride

  • MDL number MFCD09026838
  • Cas Number : 1187928-05-7
  • Molecular Weight : 213.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart