LIF458786958 Life Chemicals Inc.

1-(tert-butoxycarbonyl)indoline-6-carboxylic acid

  • MDL number MFCD09878909
  • Cas Number : 208772-41-2
  • Molecular Weight : 263.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart